siRNA Details
| Gene Symbol: PRR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027031.1 | XP_016882520.1 | 4 | 3760 | ||
| GenBank mRNA | AB033031.1 | - | 2 | 825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR12| Gene Symbol: PRR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027031.1 | XP_016882520.1 | 4 | 3760 | ||
| GenBank mRNA | AB033031.1 | - | 2 | 825 | ||