siRNA Details
| Gene Symbol: KIAA1211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020722.1 | NP_065773.1 | 5 | 584 | ||
| XM_005265752.2 | XP_005265809.1 | 4 | 591 | |||
| XM_011534397.2 | XP_011532699.1 | 4 | 1334 | |||
| XM_017008473.1 | XP_016863962.1 | 4 | 320 | |||
| XM_017008474.1 | XP_016863963.1 | 3 | 1198 | |||
| GenBank mRNA | AK126014.1 | - | 5 | 584 | ||
| BC144536.1 | - | 4 | 320 | 9053063 | ||
| BC150615.1 | - | 4 | 320 | 9056985 | ||
| BC171783.1 | - | 4 | 320 | 9054437 | ||

KIAA1211
Genomic Map