siRNA Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 2232 | ||
| GenBank mRNA | AB033065.2 | - | 1 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NWD2| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 2232 | ||
| GenBank mRNA | AB033065.2 | - | 1 | 500 | ||