siRNA Details
Gene Symbol: NWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 4475 | ||
GenBank mRNA | AB033065.2 | - | 1 | 2743 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144990.1 | NP_001138462.1 | 7 | 4475 | ||
GenBank mRNA | AB033065.2 | - | 1 | 2743 |