siRNA Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020768.3 | NP_065819.1 | 4 | 1741 | ||
| XM_005268492.2 | XP_005268549.1 | 3 | 2132 | |||
| XM_005268493.2 | XP_005268550.1 | 3 | 1528 | |||
| XM_006714789.2 | XP_006714852.1 | 3 | 2192 | |||
| XM_011537671.2 | XP_011535973.1 | 5 | 2027 | |||
| GenBank mRNA | AB037738.1 | - | 3 | 2203 | ||
| AK292477.1 | - | 4 | 1782 | |||
| BC111962.1 | - | 2 | 1134 | 8327430 | ||
| BC113435.1 | - | 2 | 1134 | 8322487 | ||
| CR749266.1 | - | 4 | 1742 | |||

KCTD16
Genomic Map