siRNA Details
| Gene Symbol: MARCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020814.2 | NP_065865.1 | 1 | 2174 | ||
| GenBank mRNA | AB037820.1 | - | 1 | 2174 | ||
| BC048793.1 | - | 1 | 248 | |||
| BC098448.1 | - | 1 | 603 | 6209260 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCH4| Gene Symbol: MARCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020814.2 | NP_065865.1 | 1 | 2174 | ||
| GenBank mRNA | AB037820.1 | - | 1 | 2174 | ||
| BC048793.1 | - | 1 | 248 | |||
| BC098448.1 | - | 1 | 603 | 6209260 | ||