siRNA Details
| Gene Symbol: TMEM181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020823.1 | NP_065874.1 | 14 | 1600 | ||
| XM_005267074.3 | XP_005267131.1 | 14 | 1317 | |||
| XM_011535999.1 | XP_011534301.1 | 14 | 1628 | |||
| XM_011536001.2 | XP_011534303.1 | 14 | 1808 | |||
| XM_011536002.2 | XP_011534304.1 | 14 | 1782 | |||
| GenBank mRNA | AB037844.1 | - | 14 | 1602 | ||
| BC033450.1 | - | 13 | 1323 | |||

TMEM181
Genomic Map