siRNA Details
| Gene Symbol: FAM234B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020853.1 | NP_065904.1 | 4 | 643 | ||
| XM_017019706.1 | XP_016875195.1 | 6 | 954 | |||
| GenBank mRNA | AB040900.1 | - | 1 | 50 | ||
| AK125278.1 | - | 4 | 643 | |||
| BC039267.1 | - | 4 | 632 | 5274878 | ||
| BC132772.1 | - | 4 | 740 | 40146794 | ||
| BC132774.1 | - | 4 | 740 | 40146796 | ||
| CR936647.1 | - | 4 | 634 | |||

FAM234B
Genomic Map