siRNA Details
| Gene Symbol: ZNF492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020855.2 | NP_065906.1 | 4 | 3028 | ||
| GenBank mRNA | AB040906.1 | - | 4 | 2916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF492| Gene Symbol: ZNF492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020855.2 | NP_065906.1 | 4 | 3028 | ||
| GenBank mRNA | AB040906.1 | - | 4 | 2916 | ||