siRNA Details
Gene Symbol: ZSWIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020883.1 | NP_065934.1 | 14 | 3181 | ||
XM_017001913.1 | XP_016857402.1 | 12 | 2090 | |||
GenBank mRNA | AB040944.1 | - | 14 | 3159 | ||
BC013836.1 | - | 8 | 1332 |
