siRNA Details
| Gene Symbol: KIAA1549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164665.1 | NP_001158137.1 | 8 | 3680 | ||
| NM_020910.2 | NP_065961.2 | 8 | 3680 | |||
| XM_011516442.2 | XP_011514744.1 | 9 | 3907 | |||
| GenBank mRNA | AB046769.2 | - | 7 | 3424 | ||
| AM989468.1 | - | 8 | 3680 | |||
| AM989469.1 | - | 8 | 3680 | |||
| BC038232.1 | - | 3 | 261 | |||

KIAA1549
Genomic Map