siRNA Details
| Gene Symbol: SFMBT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018039.1 | NP_001018049.1 | 2 | 104 | ||
| NM_001029880.2 | NP_001025051.1 | 2 | 206 | |||
| XM_005252551.2 | XP_005252608.1 | 2 | 178 | |||
| XM_011519611.1 | XP_011517913.1 | 2 | 205 | |||
| XM_017016466.1 | XP_016871955.1 | 2 | 107 | |||
| GenBank mRNA | AB046837.1 | - | 2 | 104 | ||
| AK090887.1 | - | 2 | 212 | |||
| BC144327.1 | - | 2 | 171 | |||
| BC152430.1 | - | 2 | 104 | 8862546 | ||
| DA152101.1 | - | 2 | 214 | |||

SFMBT2
Genomic Map