siRNA Details
| Gene Symbol: WFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282466.1 | NP_001269395.1 | 1 | 368 | ||
| NM_001282467.1 | NP_001269396.1 | 1 | 368 | |||
| NM_021197.3 | NP_067020.2 | 1 | 368 | |||
| XM_005256079.3 | XP_005256136.1 | 1 | 363 | |||
| GenBank mRNA | AF169631.1 | - | 1 | 256 | ||
| AF302109.1 | - | 1 | 279 | |||
| BC029159.1 | - | 1 | 174 | 5186156 | ||
| CD519223.1 | - | 1 | 282 | |||

WFDC1
Genomic Map