siRNA Details
| Gene Symbol: WFDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282466.1 | NP_001269395.1 | 3 | 627 | ||
| NM_001282467.1 | NP_001269396.1 | 3 | 627 | |||
| NM_021197.3 | NP_067020.2 | 3 | 627 | |||
| GenBank mRNA | AF169631.1 | - | 3 | 515 | ||
| AF302109.1 | - | 3 | 538 | |||
| AK075061.1 | - | 3 | 487 | |||
| AL713785.1 | - | 1 | 2222 | |||
| BC029159.1 | - | 3 | 433 | 5186156 | ||
| BQ670532.1 | - | 3 | 250 | |||
| BQ671964.1 | - | 3 | 250 | |||
| CD519223.1 | - | 3 | 541 | |||

WFDC1
Genomic Map