siRNA Details
| Gene Symbol: LY6G5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021221.2 | NP_067044.2 | 3 | 653 | ||
| GenBank mRNA | AK289364.1 | - | 3 | 1437 | ||
| CF264683.1 | - | 4 | 686 | |||
| HE864448.1 | - | 2 | 271 | |||
| HE864449.1 | - | 2 | 435 | |||
| HE864450.1 | - | 3 | 673 | |||
| HE864451.1 | - | 2 | 820 | |||
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HE864453.1 | - | 4 | 515 | ||
| HE864454.1 | - | 4 | 543 | |||
| HE864455.1 | - | 4 | 545 | |||
| HE864459.1 | - | 4 | 1086 | |||

LY6G5B
Genomic Map