siRNA Details
| Gene Symbol: LY6G6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021246.2 | NP_067069.2 | 1, 2 | 56 | ||
| GenBank mRNA | AJ315535.1 | - | 1, 2 | 56 | ||
| AJ315537.1 | - | 1, 2 | 56 | |||
| BC125138.1 | - | 1, 2 | 60 | 40119583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY6G6D| Gene Symbol: LY6G6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021246.2 | NP_067069.2 | 1, 2 | 56 | ||
| GenBank mRNA | AJ315535.1 | - | 1, 2 | 56 | ||
| AJ315537.1 | - | 1, 2 | 56 | |||
| BC125138.1 | - | 1, 2 | 60 | 40119583 | ||