siRNA Details
| Gene Symbol: SNX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021249.3 | NP_067072.3 | 7 | 825 | ||
| NM_152233.2 | NP_689419.2 | 8 | 720 | |||
| XM_006720224.1 | XP_006720287.1 | 7 | 607 | |||
| GenBank mRNA | AF109364.1 | - | 3 | 203 | ||
| AF121856.1 | - | 8 | 711 | |||
| AK021456.1 | - | 3 | 207 | |||
| AK292250.1 | - | 8 | 703 | |||
| AK296213.1 | - | 9 | 848 | |||
| AK308464.1 | - | 9 | 848 | |||
| BC001798.1 | - | 7 | 825 | 3354467 | ||
| BI870378.1 | - | 2 | 101 | |||
| U83194.1 | - | 8 | 688 | |||

SNX6
Genomic Map