siRNA Details
| Gene Symbol: PBOV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021635.2 | NP_067648.1 | 1 | 206 | ||
| GenBank mRNA | AF189269.1 | - | 1 | 209 | ||
| AF189270.1 | - | 1 | 209 | |||
| BC069109.1 | - | 1 | 206 | 7216926 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PBOV1| Gene Symbol: PBOV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021635.2 | NP_067648.1 | 1 | 206 | ||
| GenBank mRNA | AF189269.1 | - | 1 | 209 | ||
| AF189270.1 | - | 1 | 209 | |||
| BC069109.1 | - | 1 | 206 | 7216926 | ||