siRNA Details
| Gene Symbol: CDH26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021810.4 | NP_068582.2 | 5, 6 | 491 | ||
| NM_177980.2 | NP_817089.1 | 17, 18 | 2594 | |||
| XM_017027998.1 | XP_016883487.1 | 17, 18 | 3261 | |||
| XM_017027999.1 | XP_016883488.1 | 16, 17 | 3192 | |||
| XM_017028001.1 | XP_016883490.1 | 15, 16 | 3138 | |||
| GenBank mRNA | AK055202.1 | - | 17, 18 | 2583 | ||
| AK074477.1 | - | 5, 6 | 479 | |||
| BC062570.1 | - | 3, 4 | 368 | |||
| BC136306.1 | - | 17, 18 | 2577 | 9020293 | ||
| BC143931.1 | - | 17, 18 | 2577 | |||

CDH26
Genomic Map