siRNA Details
| Gene Symbol: NYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022567.2 | NP_072089.1 | 2 | 1129 | ||
| XM_017029709.1 | XP_016885198.1 | 3 | 797 | |||
| GenBank mRNA | AF254868.1 | - | 3 | 797 | ||
| AJ278865.1 | - | 2 | 1155 | |||
| BC112242.1 | - | 2 | 743 | 8327710 | ||

NYX
Genomic Map