siRNA Details
| Gene Symbol: MS4A6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001247999.1 | NP_001234928.1 | 7 | 1182 | ||
| NM_022349.3 | NP_071744.2 | 6 | 993 | |||
| NM_152851.2 | NP_690590.1 | 6 | 889 | |||
| NM_152852.2 | NP_690591.1 | 7 | 1118 | |||
| XM_005274177.2 | XP_005274234.1 | 7 | 1177 | |||
| XM_006718658.2 | XP_006718721.1 | 7 | 1052 | |||
| XM_006718660.2 | XP_006718723.1 | 6 | 917 | |||
| XM_006718661.2 | XP_006718724.1 | 6 | 917 | |||
| XM_017018123.1 | XP_016873612.1 | 5 | 653 | |||
| XM_017018124.1 | XP_016873613.1 | 5 | 653 | |||
| GenBank mRNA | AB013104.1 | - | 6 | 597 | ||
| AF142409.1 | - | 6 | 721 | |||
| AF212240.1 | - | 6 | 862 | |||
| AF237908.1 | - | 6 | 863 | |||
| AF253977.1 | - | 6 | 866 | |||
| AF261136.1 | - | 6 | 863 | |||
| AF286866.1 | - | 5 | 641 | |||
| AF350502.1 | - | 6 | 710 | |||
| AF350503.1 | - | 6 | 710 | |||
| AF354930.1 | - | 6 | 863 | |||
| AK291870.1 | - | 7 | 1118 | |||
| AK300725.1 | - | 6 | 727 | |||
| AK301387.1 | - | 5 | 553 | |||
| BC022854.1 | - | 6 | 732 | 3934920 | ||
| BC108728.1 | - | 6 | 766 | 6196213 | ||

MS4A6A
Genomic Map