siRNA Details
| Gene Symbol: MOSPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040097.1 | NP_001035186.1 | 5 | 828 | ||
| NM_001040098.1 | NP_001035187.1 | 5 | 744 | |||
| NM_001040099.1 | NP_001035188.1 | 4 | 897 | |||
| NM_023948.4 | NP_076438.1 | 4 | 927 | |||
| XM_005250529.4 | XP_005250586.1 | 5 | 749 | |||
| XM_005250531.3 | XP_005250588.1 | 5 | 774 | |||
| GenBank mRNA | BC005042.1 | - | 4 | 856 | 3689026 | |
| BC011653.2 | - | 4 | 908 | 4299899 | ||
| BC028475.2 | - | 4 | 832 | |||

MOSPD3
Genomic Map