siRNA Details
| Gene Symbol: IL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022789.3 | NP_073626.1 | 2 | 742 | ||
| NM_172314.1 | NP_758525.1 | 3 | 594 | |||
| GenBank mRNA | AF305200.1 | - | 2 | 742 | ||
| AF461739.1 | - | 3 | 594 | |||
| AY359127.1 | - | 2 | 742 | |||
| BC069565.1 | - | 2 | 559 | 7262228 | ||
| BC104929.1 | - | 3 | 484 | 8143932 | ||
| BC104931.1 | - | 3 | 484 | 8143934 | ||

IL25
Genomic Map