siRNA Details
| Gene Symbol: MARCKSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023009.6 | NP_075385.1 | 2 | 556 | ||
| NR_052852.1 | 2 | 331 | ||||
| GenBank mRNA | AL713653.1 | - | 2 | 539 | ||
| AY007131.1 | - | 2 | 539 | |||
| BC007904.2 | - | 2 | 541 | 4138092 | ||
| BC066915.1 | - | 2 | 550 | 5259353 | ||
| DT218825.1 | - | 2 | 323 | |||
| X70326.1 | - | 2 | 363 | |||

MARCKSL1
Genomic Map