siRNA Details
| Gene Symbol: RSRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023012.5 | NP_075388.2 | 8 | 1127 | ||
| NR_036434.1 | 9 | 1208 | ||||
| NR_036435.1 | 9 | 1227 | ||||
| NR_036436.1 | 8 | 1149 | ||||
| XM_005253601.1 | XP_005253658.1 | 8 | 1130 | |||
| XM_005253602.2 | XP_005253659.1 | 8 | 1101 | |||
| XM_005253604.1 | XP_005253661.1 | 9 | 1212 | |||
| XM_011538688.1 | XP_011536990.1 | 8 | 1130 | |||
| XM_011538689.1 | XP_011536991.1 | 11 | 1385 | |||
| XM_017019830.1 | XP_016875319.1 | 8 | 1127 | |||
| XM_017019831.1 | XP_016875320.1 | 9 | 1207 | |||
| XM_017019832.1 | XP_016875321.1 | 11 | 1363 | |||
| XM_017019833.1 | XP_016875322.1 | 9 | 1154 | |||
| GenBank mRNA | AB212664.1 | - | 8 | 1113 | ||
| AF161432.1 | - | 4 | 605 | |||
| AK001883.1 | - | 8 | 1087 | |||
| AK023985.1 | - | 5 | 717 | |||
| AK056256.1 | - | 8 | 1087 | |||
| BC008684.1 | - | 8 | 1037 | 3853928 | ||
| BC067773.1 | - | 8 | 1097 | 5301415 | ||
| BX640711.1 | - | 8 | 1073 | |||
| BX647884.1 | - | 8 | 1116 | |||

RSRC2
Genomic Map