siRNA Details
| Gene Symbol: ZSWIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023072.2 | NP_075560.2 | 12 | 2234 | ||
| XM_005260023.3 | XP_005260080.1 | 13 | 2396 | |||
| XM_017027153.1 | XP_016882642.1 | 13 | 2667 | |||
| XM_017027154.1 | XP_016882643.1 | 13 | 2599 | |||
| XM_017027155.1 | XP_016882644.1 | 12 | 2500 | |||
| XM_017027156.1 | XP_016882645.1 | 12 | 2171 | |||
| GenBank mRNA | AK022283.1 | - | 1 | 17 | ||
| AK024452.1 | - | 12 | 2234 | |||
| AK299057.1 | - | 11 | 1772 | |||
| BG764264.1 | - | 4 | 436 | |||

ZSWIM4
Genomic Map