siRNA Details
| Gene Symbol: UNKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193388.3 | NP_001180317.2 | 13 | 1710 | ||
| NM_001193389.1 | NP_001180318.1 | 4 | 568 | |||
| NM_001276414.1 | NP_001263343.1 | 4 | 764 | |||
| XM_005255505.1 | XP_005255562.1 | 13 | 1739 | |||
| XM_011522610.1 | XP_011520912.1 | 14 | 1799 | |||
| XM_011522611.1 | XP_011520913.1 | 14 | 1799 | |||
| XM_011522612.1 | XP_011520914.1 | 13 | 1567 | |||
| XM_017023568.1 | XP_016879057.1 | 4 | 565 | |||
| XM_017023569.1 | XP_016879058.1 | 4 | 680 | |||
| GenBank mRNA | AK022685.1 | - | 2 | 299 | ||
| AK027013.1 | - | 4 | 569 | |||
| AK124977.1 | - | 3 | 844 | |||
| AM944365.1 | - | 13 | 1672 | |||
| BC000150.1 | - | 4 | 726 | 2900118 | ||
| DA558634.1 | - | 4 | 489 | |||

UNKL
Genomic Map