siRNA Details
| Gene Symbol: DLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286655.1 | NP_001273584.1 | 6 | 1201 | ||
| NM_001286656.1 | NP_001273585.1 | 6 | 878 | |||
| NM_023932.3 | NP_076421.2 | 6 | 896 | |||
| NM_206539.2 | NP_996262.1 | 6 | 1312 | |||
| XM_005249308.4 | XP_005249365.1 | 5 | 2104 | |||
| XM_011514823.2 | XP_011513125.1 | 5 | 1465 | |||
| XM_017011204.1 | XP_016866693.1 | 3 | 914 | |||
| GenBank mRNA | AK055380.1 | - | 6 | 1312 | ||
| AY358126.1 | - | 6 | 762 | |||
| BC000230.1 | - | 6 | 595 | 3350499 | ||
| BC006425.1 | - | 6 | 595 | 3349698 | ||
| BC110320.1 | - | 6 | 864 | 5495458 | ||

DLK2
Genomic Map