siRNA Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023934.3 | NP_076423.2 | 1 | 184 | ||
| GenBank mRNA | AF267862.1 | - | 1 | 215 | ||
| AF320778.1 | - | 1 | 147 | |||
| AJ272054.1 | - | 1 | 141 | |||
| AK313143.1 | - | 1 | 126 | |||
| AY032594.1 | - | 1 | 34 | |||
| BC000255.1 | - | 1 | 131 | 3355861 | ||
| BC072685.1 | - | 1 | 89 | 6502640 | ||
| BC108657.1 | - | 1 | 40 | 5752287 | ||
| BI463291.1 | - | 1 | 189 | |||

FUNDC2
Genomic Map