siRNA Details
| Gene Symbol: LRRC61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142928.1 | NP_001136400.1 | 3 | 1162 | ||
| NM_023942.2 | NP_076431.1 | 2 | 992 | |||
| XM_006716095.2 | XP_006716158.1 | 3 | 1382 | |||
| GenBank mRNA | AK025869.1 | - | 2 | 664 | ||
| AK055954.1 | - | 3 | 1078 | |||
| AK093915.1 | - | 1 | 1712 | |||
| AK098061.1 | - | 3 | 1168 | |||
| BC001354.1 | - | 2 | 691 | 3163787 | ||

LRRC61
Genomic Map