siRNA Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024026.4 | NP_076931.1 | 2 | 614 | ||
| XM_017020740.1 | XP_016876229.1 | 2 | 248 | |||
| GenBank mRNA | AB049957.1 | - | 1 | 136 | ||
| BC023616.2 | - | 2 | 170 | 4645641 | ||
| BC068492.1 | - | 2 | 180 | 4801279 | ||
| BG773624.1 | - | 2 | 203 | |||
| BG778041.1 | - | 2 | 186 | |||
| BI601227.1 | - | 2 | 156 | |||

MRPL57
Genomic Map