siRNA Details
| Gene Symbol: CUEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024040.2 | NP_076945.2 | 2, 3 | 226 | ||
| GenBank mRNA | AB025425.1 | - | 1, 2 | 75 | ||
| AB232358.1 | - | 1, 2 | 75 | |||
| BC000262.1 | - | 2, 3 | 156 | 3353174 | ||
| BM756951.1 | - | 2, 3 | 226 | |||
| CX165866.1 | - | 2, 3 | 110 | |||

CUEDC2
Genomic Map