siRNA Details
| Gene Symbol: SMIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024104.3 | NP_077009.2 | 5 | 332 | ||
| GenBank mRNA | AK025602.1 | - | 5 | 289 | ||
| AL702263.1 | - | 5 | 333 | |||
| BC001680.2 | - | 2 | 104 | |||
| BC001948.1 | - | 2 | 104 | |||
| BC069279.1 | - | 6 | 662 | |||
| BX648276.1 | - | 4 | 229 | |||

SMIM7
Genomic Map