siRNA Details
| Gene Symbol: CARD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257970.1 | NP_001244899.1 | 5 | 900 | ||
| NM_024110.4 | NP_077015.2 | 5 | 900 | |||
| NM_052819.2 | NP_438170.1 | 2 | 181 | |||
| NR_047566.1 | 6 | 965 | ||||
| XM_011525212.1 | XP_011523514.1 | 7 | 1421 | |||
| XM_011525213.1 | XP_011523515.1 | 7 | 1163 | |||
| XM_011525215.1 | XP_011523517.1 | 6 | 1223 | |||
| XM_011525216.1 | XP_011523518.1 | 6 | 923 | |||
| XM_011525217.1 | XP_011523519.1 | 6 | 979 | |||
| XM_011525218.2 | XP_011523520.1 | 6 | 962 | |||
| XM_017025057.1 | XP_016880546.1 | 7 | 1421 | |||
| XM_017025058.1 | XP_016880547.1 | 7 | 1421 | |||
| GenBank mRNA | AF322642.1 | - | 5 | 938 | ||
| AK091123.1 | - | 6 | 923 | |||
| AK296762.1 | - | 2 | 121 | |||
| AK309299.1 | - | 1 | 211 | |||
| AX746790.1 | - | 6 | 923 | |||
| AY032927.1 | - | 4 | 735 | |||
| BC001326.1 | - | 2 | 181 | |||
| BC018142.1 | - | 5 | 909 | 3847282 | ||
| EU652409.1 | - | 4 | 732 | |||

CARD14
Genomic Map