siRNA Details
| Gene Symbol: NOL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024313.2 | NP_077289.1 | 2, 3 | 248 | ||
| GenBank mRNA | AK057139.1 | - | 2, 3 | 231 | ||
| AK294700.1 | - | 2, 3 | 231 | |||
| AK310084.1 | - | 2, 3 | 224 | |||
| AK316443.1 | - | 2, 3 | 231 | |||
| AL160132.1 | - | 2, 3 | 214 | |||
| BC002808.1 | - | 2, 3 | 225 | 3634584 | ||
| BE241845.1 | - | 2, 3 | 249 | |||
| CR456463.1 | - | 2, 3 | 208 | |||

NOL12
Genomic Map