siRNA Details
Gene Symbol: LILRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003061.2 | 4 | 1206 | |||
GenBank mRNA | BC131616.1 | - | 4 | 1206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LILRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003061.2 | 4 | 1206 | |||
GenBank mRNA | BC131616.1 | - | 4 | 1206 |