siRNA Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016531.5 | NP_057615.3 | 1 | 263 | ||
| XM_017008279.1 | XP_016863768.1 | 1 | 233 | |||
| GenBank mRNA | AF285837.1 | - | 1 | 90 | ||
| AK313459.1 | - | 1 | 233 | |||
| BC030662.1 | - | 1 | 238 | 5262512 | ||
| BC051687.1 | - | 1 | 235 | 5303520 | ||
| DB246277.1 | - | 1 | 266 | |||
| Gene Symbol: KLF3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026804.1 | 1 | 194 | |||
| GenBank mRNA | AK023259.1 | - | 1 | 194 | ||

KLF3
Genomic Map