siRNA Details
| Gene Symbol: ADGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024694.3 | NP_078970.3 | 6 | 720 | ||
| XM_006715566.3 | XP_006715629.1 | 6 | 755 | |||
| XM_011536125.2 | XP_011534427.1 | 6 | 706 | |||
| XM_011536126.1 | XP_011534428.1 | 6 | 755 | |||
| XM_011536127.2 | XP_011534429.1 | 6 | 755 | |||
| XM_011536128.2 | XP_011534430.1 | 6 | 755 | |||
| XM_011536130.2 | XP_011534432.1 | 7 | 654 | |||
| XM_017011314.1 | XP_016866803.1 | 6 | 755 | |||
| XM_017011315.1 | XP_016866804.1 | 6 | 755 | |||
| GenBank mRNA | AK097570.1 | - | 6 | 720 | ||

ADGB
Genomic Map