siRNA Details
| Gene Symbol: CCDC82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318736.1 | NP_001305665.1 | 10 | 1811 | ||
| NM_024725.3 | NP_079001.2 | 9 | 1676 | |||
| XM_005274292.3 | XP_005274349.1 | 7 | 1556 | |||
| XM_006718911.3 | XP_006718974.1 | 10 | 1628 | |||
| XM_011542984.2 | XP_011541286.1 | 9 | 2340 | |||
| XM_011542987.2 | XP_011541289.1 | 10 | 1772 | |||
| XM_011542988.2 | XP_011541290.1 | 8 | 1648 | |||
| XM_017018305.1 | XP_016873794.1 | 8 | 3421 | |||
| XM_017018306.1 | XP_016873795.1 | 9 | 3505 | |||
| XM_017018307.1 | XP_016873796.1 | 10 | 2147 | |||
| XM_017018308.1 | XP_016873797.1 | 7 | 3381 | |||
| XM_017018309.1 | XP_016873798.1 | 7 | 1516 | |||
| XM_017018310.1 | XP_016873799.1 | 7 | 1557 | |||
| XM_017018311.1 | XP_016873800.1 | 9 | 3352 | |||
| XM_017018312.1 | XP_016873801.1 | 10 | 2271 | |||
| GenBank mRNA | AF245436.1 | - | 9 | 1664 | ||
| AK027171.1 | - | 6 | 881 | |||
| AK056790.1 | - | 9 | 1669 | |||
| AK074306.1 | - | 10 | 1804 | |||
| AK314669.1 | - | 6 | 1705 | |||
| BC033726.2 | - | 6 | 1477 | 5001231 | ||

CCDC82
Genomic Map