siRNA Details
| Gene Symbol: MAP6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024871.2 | NP_079147.1 | 1 | 444 | ||
| GenBank mRNA | AK022810.1 | - | 1 | 416 | ||
| BC006434.1 | - | 1 | 409 | 3937613 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP6D1| Gene Symbol: MAP6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024871.2 | NP_079147.1 | 1 | 444 | ||
| GenBank mRNA | AK022810.1 | - | 1 | 416 | ||
| BC006434.1 | - | 1 | 409 | 3937613 | ||