siRNA Details
| Gene Symbol: C10orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024886.2 | NP_079162.1 | 1 | 87 | ||
| GenBank mRNA | AK024342.1 | - | 1 | 87 | ||
| BC126459.1 | - | 1 | 32 | 8992175 | ||
| Gene Symbol: RPARP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038937.1 | 3 | 435 | |||
| GenBank mRNA | BC089416.1 | - | 3 | 399 | ||

C10orf95
Genomic Map