siRNA Details
| Gene Symbol: ZNF442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030824.2 | NP_110451.1 | 6 | 2285 | ||
| XM_006722908.3 | XP_006722971.1 | 5 | 1765 | |||
| XM_017027316.1 | XP_016882805.1 | 4 | 1713 | |||
| XM_017027317.1 | XP_016882806.1 | 4 | 1615 | |||
| XM_017027318.1 | XP_016882807.1 | 4 | 2218 | |||
| GenBank mRNA | AK024418.1 | - | 6 | 2257 | ||
| AK295923.1 | - | 3 | 1578 | |||

ZNF442
Genomic Map