siRNA Details
Gene Symbol: ZNF703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025069.2 | NP_079345.1 | 2 | 1224 | ||
GenBank mRNA | AK024361.1 | - | 2 | 1192 | ||
BC032534.1 | - | 2 | 1153 | 5527569 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025069.2 | NP_079345.1 | 2 | 1224 | ||
GenBank mRNA | AK024361.1 | - | 2 | 1192 | ||
BC032534.1 | - | 2 | 1153 | 5527569 |