siRNA Details
Gene Symbol: ZNF703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025069.2 | NP_079345.1 | 1, 2 | 474 | ||
GenBank mRNA | AK024361.1 | - | 1, 2 | 442 | ||
BC032534.1 | - | 1, 2 | 403 | 5527569 | ||
BC084581.1 | - | 1, 2 | 436 | |||
DA936465.1 | - | 1, 2 | 476 |
