siRNA Details
| Gene Symbol: PQLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146345.1 | NP_001139817.1 | 4 | 614 | ||
| NM_025078.4 | NP_079354.2 | 5 | 668 | |||
| XM_011526188.1 | XP_011524490.1 | 3 | 279 | |||
| XM_017025997.1 | XP_016881486.1 | 6 | 755 | |||
| XM_017025998.1 | XP_016881487.1 | 5 | 701 | |||
| XM_017025999.1 | XP_016881488.1 | 6 | 755 | |||
| XM_017026001.1 | XP_016881490.1 | 4 | 685 | |||
| XM_017026005.1 | XP_016881494.1 | 4 | 403 | |||
| GenBank mRNA | AK026031.1 | - | 2 | 244 | ||
| AK123870.1 | - | 5 | 539 | |||
| AK126188.1 | - | 4 | 1239 | |||
| BC030140.2 | - | 5 | 632 | 4761671 | ||

PQLC1
Genomic Map