siRNA Details
| Gene Symbol: C16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025108.2 | NP_079384.2 | 9 | 1169 | ||
| XM_011522667.1 | XP_011520969.1 | 8 | 1073 | |||
| GenBank mRNA | AK023971.1 | - | 8 | 1250 | ||
| AK093788.1 | - | 8 | 1407 | |||
| AK128408.1 | - | 7 | 1999 | |||
| AK301929.1 | - | 9 | 1165 | |||
| BC008882.2 | - | 6 | 706 | |||
| BC018719.1 | - | 9 | 1157 | 4868952 | ||
| CR457319.1 | - | 6 | 637 | |||

C16orf59
Genomic Map