siRNA Details
| Gene Symbol: C16orf59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025108.2 | NP_079384.2 | 10 | 1293 | ||
| XM_011522667.1 | XP_011520969.1 | 9 | 1197 | |||
| GenBank mRNA | AK023971.1 | - | 9 | 1374 | ||
| AK093788.1 | - | 8 | 1823 | |||
| AK128408.1 | - | 7 | 2415 | |||
| AK301929.1 | - | 10 | 1289 | |||
| BC018719.1 | - | 10 | 1281 | 4868952 | ||
| CR457319.1 | - | 7 | 761 | |||

C16orf59
Genomic Map