siRNA Details
| Gene Symbol: TM2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025141.3 | NP_079417.2 | 5 | 549 | ||
| NM_078474.2 | NP_510883.2 | 6 | 627 | |||
| GenBank mRNA | AF353992.1 | - | 5 | 524 | ||
| AK026257.1 | - | 5 | 539 | |||
| AK094955.1 | - | 4 | 959 | |||
| AK315584.1 | - | 6 | 603 | |||
| AL512689.1 | - | 2 | 1385 | |||
| BC006150.1 | - | 6 | 599 | 3051281 | ||
| BC008873.2 | - | 5 | 522 | 4137334 | ||

TM2D3
Genomic Map