siRNA Details
| Gene Symbol: TM2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025141.3 | NP_079417.2 | 5 | 648 | ||
| NM_078474.2 | NP_510883.2 | 6 | 726 | |||
| GenBank mRNA | AF353992.1 | - | 5 | 623 | ||
| AK026257.1 | - | 5 | 638 | |||
| AK094955.1 | - | 4 | 1058 | |||
| AK315584.1 | - | 6 | 702 | |||
| AL512689.1 | - | 2 | 1484 | |||
| BC006150.1 | - | 6 | 698 | 3051281 | ||
| BC008873.2 | - | 5 | 621 | 4137334 | ||

TM2D3
Genomic Map