siRNA Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025208.4 | NP_079484.1 | 1 | 455 | ||
| NM_033135.3 | NP_149126.1 | 1 | 455 | |||
| GenBank mRNA | AB033832.1 | - | 1 | 239 | ||
| AF113216.1 | - | 1 | 189 | |||
| AF335584.1 | - | 1 | 264 | |||
| AF336376.1 | - | 1 | 258 | |||
| AK292801.1 | - | 1 | 263 | |||
| AY027517.1 | - | 1 | 175 | |||
| AY027518.1 | - | 1 | 175 | |||
| AY359116.1 | - | 1 | 114 | |||
| BC030645.2 | - | 1 | 535 | 4824526 | ||
| BG722948.1 | - | 1 | 326 | |||

PDGFD
Genomic Map